{[5-(Methoxymethyl)-1,3,4-thiadiazol-2-yl]amino}(oxo)acetic acid

Supplier: Matrix Scientific

080224-500MG 080224-5G
102668-512EA 387.69 USD
102668-512 102668-516
{[5-(Methoxymethyl)-1,3,4-thiadiazol-2-yl]amino}(oxo)acetic acid
{[5-(Methoxymethyl)-1,3,4-thiadiazol-2-yl]amino}(oxo)acetic acid

Hazard: Irritant

Formula: C₆H₇N₃O₄S
MDL Number: MFCD15181810
CAS Number: 1158341-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR