[(5-Isobutyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid

Supplier: Matrix Scientific

080222-500MG 080222-5G
102668-500EA 387.69 USD
102668-500 102668-504
[(5-Isobutyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid
[(5-Isobutyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₈H₁₁N₃O₃S
MDL Number: MFCD15181806
CAS Number: 83244-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR