4-[(5-Mercapto-1,3,4-thiadiazol-2-yl)amino]-5-oxo-1-(2-phenylethyl)-2,5-dihydro-1H-pyrrole-3-carboxylic acid

Supplier: Matrix Scientific

077909-500MG 077909-5G
102668-286EA 193.85 USD
102668-286 102668-290
4-[(5-Mercapto-1,3,4-thiadiazol-2-yl)amino]-5-oxo-1-(2-phenylethyl)-2,5-dihydro-1H-pyrrole-3-carboxylic acid
4-[(5-Mercapto-1,3,4-thiadiazol-2-yl)amino]-5-oxo-1-(2-phenylethyl)-2,5-dihydro-1H-pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₄N₄O₃S₂
MDL Number: MFCD22056563
CAS Number: 1111018-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR