1-tert-Butyl-4-(1-chloroethyl)benzene

Supplier: Matrix Scientific

077904-500MG 077904-5G
102668-266EA 387.69 USD
102668-266 102668-270
1-tert-Butyl-4-(1-chloroethyl)benzene
1-tert-Butyl-4-(1-chloroethyl)benzene

Hazard: Irritant

Formula: C₁₂H₁₇Cl
MDL Number: MFCD11186417
CAS Number: 13372-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR