Methyl-1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1H-pyrrole-3-carboxylate

Supplier: Matrix Scientific

077901-500MG 077901-5G
102668-258EA 387.69 USD
102668-258 102668-262
Methyl-1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1H-pyrrole-3-carboxylate
Methyl-1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1H-pyrrole-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₄
MDL Number: MFCD22056562
CAS Number: 96124-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR