1-(4-Methylpyrimidin-2-yl)-1,4-diazepane hydrochloride
Supplier: Matrix Scientific
077894-500MG
077894-5G
102668-230EA
387.69
USD
102668-230
102668-234
1-(4-Methylpyrimidin-2-yl)-1,4-diazepane hydrochloride
1-(4-Methylpyrimidin-2-yl)-1,4-diazepane hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₇ClN₄ |
MDL Number:
MFCD22123310 CAS Number: 1401425-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...