Methyl-N-methylalaninate hydrochloride

Supplier: Matrix Scientific

077881-500MG 077881-5G
102668-172EA 516.92 USD
102668-172 102668-176
Methyl-N-methylalaninate hydrochloride
Methyl-N-methylalaninate hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO₂
MDL Number: MFCD18432495
CAS Number: 114079-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR