Ethyl morpholin-4-yl(oxo)acetate

Supplier: Matrix Scientific

077873-500MG 077873-5G
102668-136EA 387.69 USD
102668-136 102668-140
Ethyl morpholin-4-yl(oxo)acetate
Ethyl morpholin-4-yl(oxo)acetate

Hazard: Irritant

Formula: C₈H₁₃NO₄
MDL Number: MFCD02859949
CAS Number: 20943-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR