Ethyl-1-[(dimethylamino)carbonyl]piperidine-3-carboxylate

Supplier: Matrix Scientific

077865-500MG 077865-5G
102668-098EA 387.69 USD
102668-098 102668-102
Ethyl-1-[(dimethylamino)carbonyl]piperidine-3-carboxylate
Ethyl-1-[(dimethylamino)carbonyl]piperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₃
MDL Number: MFCD05702606
CAS Number: 831200-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR