2-(Methylamino)butanoic acid hydrochloride
Supplier: Matrix Scientific
077860-500MG
077860-5G
102668-074EA
516.92
USD
102668-074
102668-078
2-(Methylamino)butanoic acid hydrochloride
2-(Methylamino)butanoic acid hydrochloride
Hazard: Irritant
Formula:
C₅H₁₂ClNO₂ |
MDL Number:
MFCD13196063 CAS Number: 1214138-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...