1-(Difluoromethyl)-4-iodo-5-methyl-1H-pyrazole

Supplier: Matrix Scientific

077829-500MG 077829-5G
102667-950EA 516.92 USD
102667-950 102667-954
1-(Difluoromethyl)-4-iodo-5-methyl-1H-pyrazole
1-(Difluoromethyl)-4-iodo-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₅F₂IN₂
MDL Number: MFCD21364399
CAS Number: 1354705-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR