3-Bromo-5-cyclobutyl-1,2,4-oxadiazole
Supplier: Matrix Scientific
077822-500MG
077822-5G
102667-920EA
646.15
USD
102667-920
102667-924
3-Bromo-5-cyclobutyl-1,2,4-oxadiazole
3-Bromo-5-cyclobutyl-1,2,4-oxadiazole
Hazard: Irritant
Formula:
C₆H₇BrN₂O |
MDL Number:
MFCD21364386 CAS Number: 1359822-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...