(2-Methylprop-2-en-1-yl)hydrazine
Supplier: Matrix Scientific
077792-500MG
077792-5G
102667-774EA
646.15
USD
102667-774
102667-778
(2-Methylprop-2-en-1-yl)hydrazine
(2-Methylprop-2-en-1-yl)hydrazine
Hazard: Irritant
Formula:
C₄H₁₀N₂ |
MDL Number:
MFCD07364245 CAS Number: 146097-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...