1,1-Diethoxy-4-iodobutane
Supplier: Matrix Scientific
077790-500MG
077790-5G
102667-762EA
769.23
USD
102667-762
102667-766
1,1-Diethoxy-4-iodobutane
1,1-Diethoxy-4-iodobutane
Hazard: Irritant
Formula:
C₈H₁₇IO₂ |
MDL Number:
MFCD22056547 CAS Number: 1365964-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...