Ethyl (2E)-2-benzoyl-3-(methylamino)but-2-enoate
Supplier: Matrix Scientific
077787-500MG
077787-5G
102667-750EA
193.85
USD
102667-750
102667-754
Ethyl (2E)-2-benzoyl-3-(methylamino)but-2-enoate
Ethyl (2E)-2-benzoyl-3-(methylamino)but-2-enoate
Hazard: Irritant
Formula:
C₁₄H₁₇NO₃ |
MDL Number:
MFCD13705179 CAS Number: 82140-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...