[2-(1-Ethyl-1H-pyrazol-5-yl)-1-methylethyl]amine

Supplier: Matrix Scientific

077782-500MG 077782-5G
102667-722EA 549.23 USD
102667-722 102667-726
[2-(1-Ethyl-1H-pyrazol-5-yl)-1-methylethyl]amine
[2-(1-Ethyl-1H-pyrazol-5-yl)-1-methylethyl]amine

Hazard: Irritant

Formula: C₈H₁₅N₃
MDL Number: MFCD22056544
CAS Number: 1365964-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR