3-Ethyl-1-methyl-1H-pyrazole
Supplier: Matrix Scientific
077765-500MG
077765-5G
102667-628EA
446.15
USD
102667-628
102667-632
3-Ethyl-1-methyl-1H-pyrazole
3-Ethyl-1-methyl-1H-pyrazole
Hazard: Irritant
Formula:
C₆H₁₀N₂ |
MDL Number:
MFCD08701123 CAS Number: 30433-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...