4-Ethoxy-1-benzofuran-6-carboxylic acid
Supplier: Matrix Scientific
077740-500MG
077740-5G
102667-524EA
516.92
USD
102667-524
102667-528
4-Ethoxy-1-benzofuran-6-carboxylic acid
4-Ethoxy-1-benzofuran-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₀O₄ |
MDL Number:
MFCD14704812 CAS Number: 1239847-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...