4-(Allyloxy)-1-benzofuran-6-carboxylic acid

Supplier: Matrix Scientific

077693-1G 077693-5G
102667-494EA 646.15 USD
102667-494 102667-496
4-(Allyloxy)-1-benzofuran-6-carboxylic acid
4-(Allyloxy)-1-benzofuran-6-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₄
MDL Number: MFCD18838165
CAS Number: 1291489-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR