4-(Difluoromethoxy)-1-benzofuran-6-carboxylic acid

Supplier: Matrix Scientific

077692-500MG 077692-5G
102667-486EA 646.15 USD
102667-486 102667-490
4-(Difluoromethoxy)-1-benzofuran-6-carboxylic acid
4-(Difluoromethoxy)-1-benzofuran-6-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆F₂O₄
MDL Number: MFCD18916886
CAS Number: 1365962-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR