(2Z)-2-Chloro-3-phenylacrylic acid
Supplier: Matrix Scientific
077688-500MG
077688-5G
102667-466EA
166.15
USD
102667-466
102667-470
(2Z)-2-Chloro-3-phenylacrylic acid
(2Z)-2-Chloro-3-phenylacrylic acid
Hazard: Irritant
Formula:
C₉H₇ClO₂ Melting Pt: 137 °C |
MDL Number:
MFCD18432230 CAS Number: 705-54-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...