4-(4-Methoxyphenyl)-1,3-thiazole-2-carbaldehyde

Supplier: Matrix Scientific

077668-500MG 077668-5G
102667-376EA 775.38 USD
102667-376 102667-380
4-(4-Methoxyphenyl)-1,3-thiazole-2-carbaldehyde
4-(4-Methoxyphenyl)-1,3-thiazole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₉NO₂S
MDL Number: MFCD02665603
CAS Number: 383144-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR