Sodium-2-pyrrolidin-1-yl-1,3-thiazole-4-carboxylate

Supplier: Matrix Scientific

077656-500MG 077656-1G
102667-346EA 581.54 USD
102667-346 102667-348
Sodium-2-pyrrolidin-1-yl-1,3-thiazole-4-carboxylate
Sodium-2-pyrrolidin-1-yl-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₉N₂NaO₂S
MW: 220.23 g/mol
MDL Number: MFCD22056529
CAS Number: 1401425-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR