{[2-(4-Methylphenyl)-1,3-thiazol-5-yl]methyl}amine dihydrochloride
Supplier: Matrix Scientific
077655-500MG
077655-1G
077655-5G
102667-340EA
1130.77
USD
102667-340
102667-342
102667-344
{[2-(4-Methylphenyl)-1,3-thiazol-5-yl]methyl}amine dihydrochloride
{[2-(4-Methylphenyl)-1,3-thiazol-5-yl]methyl}amine dihydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄Cl₂N₂S |
MDL Number:
MFCD22123370 CAS Number: 1401425-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...