3-(2,5-Dimethylphenyl)-1H-pyrazole-5-carboxylic acid

Supplier: Matrix Scientific

077648-500MG 077648-1G 077648-5G
102667-304EA 387.69 USD
102667-304 102667-306 102667-308
3-(2,5-Dimethylphenyl)-1H-pyrazole-5-carboxylic acid
3-(2,5-Dimethylphenyl)-1H-pyrazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD04325788
CAS Number: 1038829-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR