[4-Hydroxy-2-(methylthio)pyrimidin-5-yl]acetic acid

Supplier: Matrix Scientific

077646-500MG 077646-1G 077646-5G
102667-292EA 387.69 USD
102667-292 102667-294 102667-296
[4-Hydroxy-2-(methylthio)pyrimidin-5-yl]acetic acid
[4-Hydroxy-2-(methylthio)pyrimidin-5-yl]acetic acid

Hazard: Irritant

Formula: C₇H₈N₂O₃S
MDL Number: MFCD19981859
CAS Number: 1087605-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR