Ethyl [2-(cyanoamino)-4-hydroxypyrimidin-5-yl]acetate
Supplier: Matrix Scientific
077644-500MG
077644-1G
102667-282EA
387.69
USD
102667-282
102667-284
Ethyl [2-(cyanoamino)-4-hydroxypyrimidin-5-yl]acetate
Ethyl [2-(cyanoamino)-4-hydroxypyrimidin-5-yl]acetate
Hazard: Irritant
Formula:
C₉H₁₀N₄O₃ |
MDL Number:
MFCD20441410 CAS Number: 1338494-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...