4-(Chloromethyl)-1,3-dimethyl-1H-pyrazole hydrochloride

Supplier: Matrix Scientific

077642-500MG 077642-1G 077642-5G
102667-272EA 646.15 USD
102667-272 102667-274 102667-276
4-(Chloromethyl)-1,3-dimethyl-1H-pyrazole hydrochloride
4-(Chloromethyl)-1,3-dimethyl-1H-pyrazole hydrochloride

Hazard: Irritant

Formula: C₆H₁₀Cl₂N₂
MDL Number: MFCD20441438
CAS Number: 1390654-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR