1-Ethyl-5-methyl-3-nitro-1H-pyrazole
Supplier: Matrix Scientific
077640-500MG
077640-1G
077640-5G
102667-260EA
516.92
USD
102667-260
102667-262
102667-264
1-Ethyl-5-methyl-3-nitro-1H-pyrazole
1-Ethyl-5-methyl-3-nitro-1H-pyrazole
Hazard: Irritant
Formula:
C₆H₉N₃O₂ |
MDL Number:
MFCD04971136 CAS Number: 1005576-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...