2,2,2-Trichloro-1-(1-isopropyl-1H-pyrazol-4-yl)ethanol
Supplier: Matrix Scientific
077636-500MG
077636-1G
102667-238EA
387.69
USD
102667-238
102667-240
2,2,2-Trichloro-1-(1-isopropyl-1H-pyrazol-4-yl)ethanol
2,2,2-Trichloro-1-(1-isopropyl-1H-pyrazol-4-yl)ethanol
Hazard: Irritant
Formula:
C₈H₁₁Cl₃N₂O |
MDL Number:
MFCD20441408 CAS Number: 1338494-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...