2,2,2-Trichloro-1-(1-ethyl-1H-pyrazol-4-yl)ethanol
Supplier: Matrix Scientific
077635-500MG
077635-1G
077635-5G
102667-232EA
387.69
USD
102667-232
102667-234
102667-236
2,2,2-Trichloro-1-(1-ethyl-1H-pyrazol-4-yl)ethanol
2,2,2-Trichloro-1-(1-ethyl-1H-pyrazol-4-yl)ethanol
Hazard: Irritant
Formula:
C₇H₉Cl₃N₂O |
MDL Number:
MFCD20441407 CAS Number: 1338495-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...