2,2,2-Trichloro-1-(1,3,5-trimethyl-1H-pyrazol-4-yl)ethanol

Supplier: Matrix Scientific

077630-500MG 077630-1G 077630-5G
102667-202EA 387.69 USD
102667-202 102667-204 102667-206
2,2,2-Trichloro-1-(1,3,5-trimethyl-1H-pyrazol-4-yl)ethanol
2,2,2-Trichloro-1-(1,3,5-trimethyl-1H-pyrazol-4-yl)ethanol

Hazard: Irritant

Formula: C₈H₁₁Cl₃N₂O
MDL Number: MFCD20441402
CAS Number: 1338494-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR