2,2,2-Trichloro-1-(1,5-dimethyl-1H-pyrazol-4-yl)ethanol
Supplier: Matrix Scientific
077629-500MG
077629-1G
077629-5G
102667-196EA
387.69
USD
102667-196
102667-198
102667-200
2,2,2-Trichloro-1-(1,5-dimethyl-1H-pyrazol-4-yl)ethanol
2,2,2-Trichloro-1-(1,5-dimethyl-1H-pyrazol-4-yl)ethanol
Hazard: Irritant
Formula:
C₇H₉Cl₃N₂O |
MDL Number:
MFCD20441401 CAS Number: 1338495-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...