2,2,2-Trichloro-1-(1,3-dimethyl-1H-pyrazol-4-yl)ethanol

Supplier: Matrix Scientific

077628-500MG 077628-1G 077628-5G
102667-190EA 387.69 USD
102667-190 102667-192 102667-194
2,2,2-Trichloro-1-(1,3-dimethyl-1H-pyrazol-4-yl)ethanol
2,2,2-Trichloro-1-(1,3-dimethyl-1H-pyrazol-4-yl)ethanol

Hazard: Irritant

Formula: C₇H₉Cl₃N₂O
MDL Number: MFCD20441400
CAS Number: 1338495-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR