[(1-Aminocyclopropyl)methyl]dimethylamine dihydrochloride
Supplier: Matrix Scientific
077627-500MG
077627-1G
077627-5G
102667-184EA
661.54
USD
102667-184
102667-186
102667-188
[(1-Aminocyclopropyl)methyl]dimethylamine dihydrochloride
[(1-Aminocyclopropyl)methyl]dimethylamine dihydrochloride
Hazard: Irritant
Formula:
C₆H₁₆Cl₂N₂ |
MDL Number:
MFCD20441459 CAS Number: 1401426-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...