2,5,6,7,8,9,10,11-Octahydro-3H-5,9:7,11-dimethano-[1,2,4]triazolo[4,3-a]azonin-3-one

Supplier: Matrix Scientific

077621-500MG 077621-1G
102667-156EA 387.69 USD
102667-156 102667-158
2,5,6,7,8,9,10,11-Octahydro-3H-5,9:7,11-dimethano-[1,2,4]triazolo[4,3-a]azonin-3-one
2,5,6,7,8,9,10,11-Octahydro-3H-5,9:7,11-dimethano-[1,2,4]triazolo[4,3-a]azonin-3-one

Hazard: Irritant

Formula: C₁₁H₁₅N₃O
MDL Number: MFCD20441398
CAS Number: 117552-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR