3,5-Bis(carboxymethyl)adamantane-1-carboxylic acid

Supplier: Matrix Scientific

077618-500MG 077618-1G
102667-140EA 492.31 USD
102667-140 102667-142
3,5-Bis(carboxymethyl)adamantane-1-carboxylic acid
3,5-Bis(carboxymethyl)adamantane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₂₀O₆
MDL Number: MFCD20441395
CAS Number: 1338494-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR