3-(Carboxymethyl)-5,7-dimethyladamantane-1-carboxylic acid

Supplier: Matrix Scientific

077617-500MG 077617-1G 077617-5G
102667-134EA 258.46 USD
102667-134 102667-136 102667-138
3-(Carboxymethyl)-5,7-dimethyladamantane-1-carboxylic acid
3-(Carboxymethyl)-5,7-dimethyladamantane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₂₂O₄
MDL Number: MFCD20441394
CAS Number: 1338494-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR