[1-(Morpholin-4-ylmethyl)cyclopropyl]amine dihydrochloride

Supplier: Matrix Scientific

077614-500MG 077614-1G 077614-5G
102667-116EA 775.38 USD
102667-116 102667-118 102667-120
[1-(Morpholin-4-ylmethyl)cyclopropyl]amine dihydrochloride
[1-(Morpholin-4-ylmethyl)cyclopropyl]amine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₈Cl₂N₂O
MW: 229.15 g/mol
MDL Number: MFCD20441462
CAS Number: 1401425-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR