[1-(Piperidin-1-ylmethyl)cyclopropyl]amine dihydrochloride

Supplier: Matrix Scientific

077613-500MG 077613-5G
102667-110EA 775.38 USD
102667-110 102667-114
[1-(Piperidin-1-ylmethyl)cyclopropyl]amine dihydrochloride
[1-(Piperidin-1-ylmethyl)cyclopropyl]amine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂
MW: 227.18 g/mol
MDL Number: MFCD18071202
CAS Number: 1255717-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR