[1-(Pyrrolidin-1-ylmethyl)cyclopropyl]amine dihydrochloride
Supplier: Matrix Scientific
077612-500MG
077612-1G
077612-5G
102667-104EA
775.38
USD
102667-104
102667-106
102667-108
[1-(Pyrrolidin-1-ylmethyl)cyclopropyl]amine dihydrochloride
[1-(Pyrrolidin-1-ylmethyl)cyclopropyl]amine dihydrochloride
Hazard: Irritant
Formula:
C₈H₁₈Cl₂N₂ |
MDL Number:
MFCD20441463 CAS Number: 1401426-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...