[1-(1-Methyl-1H-pyrazol-4-yl)cyclopropyl]amine dihydrochloride

Supplier: Matrix Scientific

077611-500MG 077611-1G 077611-5G
102667-098EA 984.62 USD
102667-098 102667-100 102667-102
[1-(1-Methyl-1H-pyrazol-4-yl)cyclopropyl]amine dihydrochloride
[1-(1-Methyl-1H-pyrazol-4-yl)cyclopropyl]amine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₃Cl₂N₃
MDL Number: MFCD20441460
CAS Number: 1401426-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR