[1-(3-Furyl)cyclopropyl]amine hydrochloride

Supplier: Matrix Scientific

077610-500MG 077610-1G 077610-5G
102667-092EA 1033.85 USD
102667-092 102667-094 102667-096
[1-(3-Furyl)cyclopropyl]amine hydrochloride
[1-(3-Furyl)cyclopropyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClNO
MDL Number: MFCD20441461
CAS Number: 1401425-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR