2-Chloro-5-fluoro-N-(3-methoxypropyl)pyrimidin-4-amine
Supplier: Matrix Scientific
077603-500MG
077603-1G
077603-5G
102667-060EA
516.92
USD
102667-060
102667-062
102667-064
2-Chloro-5-fluoro-N-(3-methoxypropyl)pyrimidin-4-amine
2-Chloro-5-fluoro-N-(3-methoxypropyl)pyrimidin-4-amine
Hazard: Irritant
Formula:
C₈H₁₁ClFN₃O MW: 219.65 g/mol |
MDL Number:
MFCD20441388 CAS Number: 1338494-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...