1-(2-Chloro-5-fluoropyrimidin-4-yl)azepane

Supplier: Matrix Scientific

077599-500MG 077599-1G 077599-5G
102667-040EA 516.92 USD
102667-040 102667-042 102667-044
1-(2-Chloro-5-fluoropyrimidin-4-yl)azepane
1-(2-Chloro-5-fluoropyrimidin-4-yl)azepane

Hazard: Irritant

Formula: C₁₀H₁₃ClFN₃
MDL Number: MFCD20441386
CAS Number: 1338495-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR