1-Ethyl-6-oxo-1,6-dihydropyrimidine-5-carbonitrile

Supplier: Matrix Scientific

077588-500MG 077588-1G
102667-014EA 516.92 USD
102667-014 102667-016
1-Ethyl-6-oxo-1,6-dihydropyrimidine-5-carbonitrile
1-Ethyl-6-oxo-1,6-dihydropyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₇H₇N₃O
MDL Number: MFCD19982743
CAS Number: 1330756-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR