2-Methyl-5-(methylsulfonyl)pyrimidin-4(3H)-one

Supplier: Matrix Scientific

077586-500MG 077586-1G 077586-5G
102667-004EA 516.92 USD
102667-004 102667-006 102667-008
2-Methyl-5-(methylsulfonyl)pyrimidin-4(3H)-one
2-Methyl-5-(methylsulfonyl)pyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₆H₈N₂O₃S
MDL Number: MFCD19982742
CAS Number: 17551-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR