4-(4H-1,2,4-Triazol-3-yl)piperidine dihydrochloride

Supplier: Matrix Scientific

077581-500MG 077581-1G 077581-5G
102666-980EA 615.38 USD
102666-980 102666-982 102666-984
4-(4H-1,2,4-Triazol-3-yl)piperidine dihydrochloride
4-(4H-1,2,4-Triazol-3-yl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₄Cl₂N₄
MDL Number: MFCD20441437
CAS Number: 1401425-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR