1-(4H-1,2,4-Triazol-3-ylmethyl)piperazine dihydrochloride
Supplier: Matrix Scientific
077580-500MG
077580-1G
077580-5G
102666-974EA
646.15
USD
102666-974
102666-976
102666-978
1-(4H-1,2,4-Triazol-3-ylmethyl)piperazine dihydrochloride
1-(4H-1,2,4-Triazol-3-ylmethyl)piperazine dihydrochloride
Hazard: Irritant
Formula:
C₇H₁₅Cl₂N₅ |
MDL Number:
MFCD20441429 CAS Number: 1401425-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...