4-(Dimethoxymethyl)-5-methylpyrimidin-2-amine

Supplier: Matrix Scientific

077579-500MG 077579-1G
102666-970EA 646.15 USD
102666-970 102666-972
4-(Dimethoxymethyl)-5-methylpyrimidin-2-amine
4-(Dimethoxymethyl)-5-methylpyrimidin-2-amine

Hazard: Irritant

Formula: C₈H₁₃N₃O₂
MDL Number: MFCD20441380
CAS Number: 1338494-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR